Phosphate-citrate Buffer (5X)
Lieferant: Thermo Fisher Scientific
J62982.AE
J62982.AEEA 63.4 EUR
J62982.AE
Phosphate-citrate Buffer (5X)
Puffer Assay-Puffer
Phosphate-Citrate Buffer (5X) is used as a substrate buffer in immunoassay procedures. It is used in SDS PAGE electrophoresis.
Contains 1M sodium phosphate and 0.5M sodium citrate.