Penicillin G Natriumsalz

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptan-2-carbonsäure Natriumsalz, Natrium-[2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptan-2-carboxylat, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6- (2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptan-2-carbonsäure Natriumsalz, 6-(2-Phenylacetamido)penicillansäure Natriumsalz, Benzylpenicillin Natriumsalz

J63032.06 J63032.14 J63032.22
J63032.06EA 33.4 EUR
J63032.06 J63032.14 J63032.22
Penicillin G Natriumsalz
Penicillin G Natriumsalz

CAS No.: 69-57-8 Molecular Formula: C16H17N2NaO4S Formula Weight: 356.37 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Beilstein Reference: 3834217 Merck Reference: 14,7094 MDL No.: MFCD00069666


Inhibits bacterial cell wall synthesis

Formel: C₁₆H₁₇N₂NaO₄S
Molecular Weight: 356.38 g/mol
Schmelzpunkt: 255
Dichte: 1.38 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00069666
CAS-Nummer: 69-57-8
EINECS: 200-710-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR