Polymyxin B-Sulfat

Lieferant: Thermo Fisher Scientific

J63074.03 J63074.06
J63074.03EA 123 EUR
J63074.03 J63074.06
Polymyxin B-Sulfat
Polymyxin B sulphate

Antibiotic for Gram-negative bacteria.


  • White to pale yellow powder


Soluble in water at 50mg/ml

Formel: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Lagertemperatur: Kühlschrank
MDL: MFCD00131991
CAS-Nummer: 1405-20-5
Merck Index: 14,7573

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR