20-Hydroxyecdysone
Lieferant: Thermo Fisher Scientific
J63091.MA
J63091.MF
J63091.MAEA
208
EUR
J63091.MA
J63091.MF
20-Hydroxyecdysone
20-Hydroxyecdysone
CAS No.: 5289-74-7
Molecular Formula: C27H44O7
Formula Weight: 480.64
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 242-244°
Solubility: Soluble in alcohol
Beilstein Reference: 1917578
MDL No.: MFCD00036740
Applications: An insect steroid hormone that induces apoptosis
Formel:
C₂₇H₄₄O₇ Schmelzpunkt: 242…244 °C |
MDL:
MFCD00036740 CAS-Nummer: 5289-74-7 Merck Index: 14,3491 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...