Triflumuron ≥98%
Lieferant: Thermo Scientific
J63112.22
J63112.22EA
319
EUR
J63112.22
Triflumuron ≥98%
Triflumuron
Purity: 98+%
CAS No.: 64628-44-0
Molecular Formula: C15H10ClF3N2O3
Formula Weight: 358.70
Physical Form: Solid
Melting Point: 198°
Beilstein Reference: 2776684
Merck Reference: 14,9678
MDL No.: MFCD00072496
Triflumuron acts as a chitin synthesis inhibitor and prevents uridine incorporation into RNA
Formel:
C₁₅H₁₀ClF₃N₂O₃ Schmelzpunkt: 198 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00072496 CAS-Nummer: 64628-44-0 EINECS: 264-980-3 Merck Index: 14,9678 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...