Enzyme storage buffer in PBS and glycerol, autoclaved

Lieferant: Thermo Fisher Scientific

J63115.AK
J63115.AKEA 60.3 EUR
J63115.AK
Enzyme storage buffer in PBS and glycerol, autoclaved
Puffer Enzympuffer

Physical Form:
Applications: For frozen storage of enzymes.

Bestellen

TECHNISCHE DATEN

Physical form Liquid
Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR