7-(2,3-Dihydroxypropyl)theophyllin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dyphylline, 7-(2,3-Dihydroxypropyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione
J63167.22
J63167.22EA
88.3
EUR
J63167.22
7-(2,3-Dihydroxypropyl)theophyllin
7-(2,3-Dihydroxypropyl)theophyllin
CAS No.: 479-18-5
Molecular Formula: C10H14N4O4
Formula Weight: 254.24
Physical Form: Powder
Appearance: White to off-white
Melting Point: 160-165°
Beilstein Reference: 284563
Merck Reference: 14,3479
MDL No.: MFCD00005759
An adenosine receptor antagonist, phosphodiesterase inhibitor and vasodilator
Formel:
C₁₀H₁₄N₄O₄ Molecular Weight: 254,24 g/mol Schmelzpunkt: 160…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005759 CAS-Nummer: 479-18-5 EINECS: 207-526-1 Merck Index: 14,3479 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...