Galaktit ≥99%
Lieferant: Thermo Scientific
Synonyme:
Dulcitol
J63206.36
J63206.36EA
238
EUR
J63206.36
Galaktit ≥99%
Galaktit
Purity: 99%
CAS No.: 608-66-2
Molecular Formula: C6H14O6
Formula Weight: 182.17
Physical Form: Powder
Density: 1.47
Appearance: White to off-white
Melting Point: 187-191°
Boiling Point: 275-280°/1mm
Beilstein Reference: 1721903
Merck Reference: 14,4332
MDL No.: MFCD00064288
Useful reagent in bacteriology
Formel:
HOH₂C(CH(OH))₄CH₂OH Molecular Weight: 182,17 g/mol Siedepunkt: 275…280 °C (1 mmHg) Schmelzpunkt: 187…191 °C Dichte: 1,47 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064288 CAS-Nummer: 608-66-2 EINECS: 210-165-2 Merck Index: 14,4332 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...