Pyrilamine maleate
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Mepyramine maleate, Anisopyradamine, Minihist, Paraminyl maleate, N1-(4-Methoxybenzyl)-N2,N2-dimethyl-N1-(pyridin-2-yl)ethane-1,2-diamine maleate, Diaminide maleate
J63247.14
J63247.14EA
132
EUR
J63247.14
Pyrilamine maleate
Pyrilamine maleate
CAS No.: 59-33-6
Molecular Formula: C17H23N3O.C4H4O4
Formula Weight: 401.46
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water at 100mg/ml
MDL No.: MFCD00069333
Applications: Selective inverse agonist for the H-1 histamine receptor
Formel:
C₂₁H₂₇N₃O₅ Molecular Weight: 401,463 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069333 CAS-Nummer: 59-33-6 EINECS: 200-422-7 Merck Index: 14,7984 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...