TAPS, 0,2 M buffer soln., pH 8,5
Lieferant: Thermo Fisher Scientific
J63268.AE
J63268.AEEA
82.1
EUR
J63268.AE
TAPS, 0,2 M buffer soln., pH 8,5
Puffer
Biologische Puffer
CAS No.: 91000-53-2
Physical Form: Liquid
MDL No.: MFCD00069903
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...