Calmidazolium chloride

Lieferant: Thermo Scientific

Synonyme: 1-(Bis(4-chlorophenyl)methyl)-3-(2-((2,4-dichlorobenzyl)oxy)-2-(2,4-dichlorophenyl)ethyl)-1H-imidazol-3-ium chloride

J63270.MA
J63270.MAEA 191 EUR
J63270.MA
Calmidazolium chloride
Calmidazolium chloride

CAS No.: 57265-65-3 Molecular Formula: C31H23Cl7N2O Formula Weight: 687.71 Physical Form: Solid Appearance: White MDL No.: MFCD00077679


Applications: Inhibits the calcium-calmodulin dependent phosphodiesterase

Formel: C₃₁H₂₃Cl₇N₂O
Molecular Weight: 687,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077679
CAS-Nummer: 57265-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR