Transformation buffer-2, pH 7,5

Lieferant: Thermo Fisher Scientific

J63293.AK
J63293.AKEA 129 EUR
J63293.AK
Transformation buffer-2, pH 7,5
Puffer Klonpuffer

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Applications: For transformation of competent cells

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR