Clindamycin phosphate

Lieferant: Thermo Scientific
Achtung

Synonyme: Clindamycin 2-Dihydrogen Phosphate, (2R,3R,4S,5R,6R)-6-((1S,2S)-2-Chloro-1-((2S,4R)-1-methyl-4-propylpyrrolidine-2-carboxamido)propyl)-4,5-dihydroxy-2-(methylthio)tetrahydro-2H-pyran-3-yl dihydrogen phosphate

J63387.MC
J63387.MCEA 288 EUR
J63387.MC
Clindamycin phosphate
Clindamycin phosphate

CAS No.: 24729-96-2 Molecular Formula: C18H34ClN2O8PS Formula Weight: 504.96 Physical Form: Solid Appearance: White Melting Point: 114° Solubility: Freely soluble in water MDL No.: MFCD07793328


Applications: Antibiotic U-28508E, effective against Gram-positive bacteria

Formel: C₁₈H₃₄ClN₂O₈PS
Molecular Weight: 504,96 g/mol
Schmelzpunkt: 114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07793328
CAS-Nummer: 24729-96-2
EINECS: 246-433-0
Merck Index: 14,2356

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR