Glipizid

Lieferant: Thermo Scientific

Synonyme: N-[4-(3-Cyclohexylureidosulphonyl)phenethyl]-5-methyl-2-pyrazinecarboxamide, 1-Cyclohexyl-3-{4-[2-(5-methylpyrazine-2-carboxamido)ethyl]phenylsulphonyl}urea, N-(4-(N-(Cyclohexylcarbamoyl)sulphamoyl)phenethyl)-5-methylpyrazine-2-carboxamide

J63398.03
J63398.03EA 104 EUR
J63398.03
Glipizid
Glipizide

CAS No.: 29094-61-9 Molecular Formula: C21H27N5O4S Formula Weight: 445.54 Physical Form: Powder Appearance: White to yellow Solubility: Soluble in methanol (1.9mg/ml) and DMSO (48mg/ml) MDL No.: MFCD00072159


Applications: An ATP-dependent potassium channel blocker

Formel: C₂₁H₂₇N₅O₄S
Molecular Weight: 445,54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00072159
CAS-Nummer: 29094-61-9
EINECS: 249-427-6
Merck Index: 14,4442

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR