Bisindolylmaleimide I

Lieferant: Thermo Scientific

Synonyme: GF 109203X

J63401.MB
J63401.MBEA 463 EUR
J63401.MB
Bisindolylmaleimide I
Bisindolylmaleimide I

Very potent and selective inhibitor of protein kinase C.


Orange crystalline powder


Applications: Very potent and selective inhibitor of protein kinase C

Formel: C₂₅H₂₄N₄O₂
Molecular Weight: 412,48 g/mol
MDL: MFCD00236428
CAS-Nummer: 133052-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR