Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt
J63427.MC
J63427.MCEA
94.6
EUR
J63427.MC
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) ≥98%
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3)
Purity: 98+%
CAS No.: 19817-92-6
Molecular Formula: C9H12N2Na3O15P3
Formula Weight: 550.09
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Beilstein Reference: 3585234
MDL No.: MFCD00044310
Formel:
C₉H₁₂N₂Na₃O₁₅P₃ Molecular Weight: 550,09 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00044310 CAS-Nummer: 19817-92-6 EINECS: 243-347-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...