Genistin ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: 5-Hydroxy-3-(4-hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

J63445.MC
J63445.MCEA 415 EUR
J63445.MC
Genistin ≥99%
Genistin

Purity: 99+% CAS No.: 529-59-9 Molecular Formula: C21H20O10 Formula Weight: 432.38 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Yellow to pale brown Melting Point: 258-259° Beilstein Reference: 64479 MDL No.: MFCD00016883


Applications: Glucoside of genistein that inhibits protein tyrosine kinase

Formel: C₂₁H₂₀O₁₀
Schmelzpunkt: 258…259 °C
MDL: MFCD00016883
CAS-Nummer: 529-59-9
Merck Index: 14,4391

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR