6-Cyan-7-nitrochinoxalin-2,3-dion 98%

Lieferant: Thermo Fisher Scientific
Achtung

J63472.MA J63472.MB J63472.MF
J63472.MAEA 144 EUR
J63472.MA J63472.MB J63472.MF
6-Cyan-7-nitrochinoxalin-2,3-dion 98%
6-Cyan-7-nitrochinoxalin-2,3-dion

Purity: 99+% CAS No.: 115066-14-3 Molecular Formula: C9H4N4O4 Formula Weight: 232.16 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 300° Solubility: Soluble in DMSO to 100mM Beilstein Reference: 5821072 MDL No.: MFCD00069232


Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist

Formel: C₉H₄N₄O₄
Schmelzpunkt: >300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00069232
CAS-Nummer: 115066-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR