Fasudil ≥98%

Lieferant: Thermo Scientific

Synonyme: HA-1077, 1-(5-Isoquinolinesulphonyl)homopiperazine

J63525.MA
J63525.MAEA 308 EUR
J63525.MA
Fasudil ≥98%
Fasudil

Purity: 98+% CAS No.: 103745-39-7 Molecular Formula: C14H17N3O2S Formula Weight: 327.83 Physical Form: Crystalline powder Appearance: White Solubility: Soluble in water or DMSO MDL No.: MFCD00866182


Applications: A potent Rho-kinase inhibitor with antivasospastic properties; inhibits vascular smooth muscle contraction by acting as an intracellular calcium antagonist

Formel: C₁₄H₁₇N₃O₂S
Molecular Weight: 327,83 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00866182
CAS-Nummer: 103745-39-7
Merck Index: 14,3942

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR