Triamcinolone acetonide ≥98%

Lieferant: Thermo Scientific
Gefahr

J63548.03
J63548.03EA 114 EUR
J63548.03
Triamcinolone acetonide ≥98%
Triamcinolone acetonide

Purity: 98+% CAS No.: 76-25-5 Molecular Formula: C24H31FO6 Formula Weight: 434.50 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 274-278° Solubility: Soluble in DMSO or ethanol. Slightly soluble in water. MDL No.: MFCD00056834


Applications: A glucocorticoid, antiasthmatic (inhalant); antiallergic (nasal)

Formel: C₂₄H₃₁FO₆
Schmelzpunkt: 274…278 °C
MDL: MFCD00056834
CAS-Nummer: 76-25-5
EINECS: 200-948-7
Merck Index: 14,9596

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR