N-Methyl-N-octanoyl-D-glucamin (MEGA-8)

Lieferant: Thermo Fisher Scientific

Synonyme: N-Octanoyl-N-methyl-D-glucamine, N-Methyl-N-((2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl)octanamide, MEGA-8

J63574.03 J63574.06
J63574.03EA 74.1 EUR
J63574.03 J63574.06
N-Methyl-N-octanoyl-D-glucamin (MEGA-8)
N-Methyl-N-octanoyl-D-glucamin (MEGA-8)

CAS No.: 85316-98-9 Molecular Formula: C15H31NO6 Formula Weight: 321.41 Physical Form: Powder Appearance: White Melting Point: 80-90° Beilstein Reference: 5816747 Optical Rotation: -18° (c=2 in water) MDL No.: MFCD00134152


Non-ionic detergent for solubilizing membrane proteins

Formel: C₁₅H₃₁NO₆
Molecular Weight: 321.4 g/mol
Schmelzpunkt: 80…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134152
CAS-Nummer: 85316-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR