N-Methyl-N-octanoyl-D-glucamin (MEGA-8)
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Octanoyl-N-methyl-D-glucamine, N-Methyl-N-((2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl)octanamide, MEGA-8
J63574.03
J63574.06
J63574.03EA
74.1
EUR
J63574.03
J63574.06
N-Methyl-N-octanoyl-D-glucamin (MEGA-8)
N-Methyl-N-octanoyl-D-glucamin (MEGA-8)
CAS No.: 85316-98-9
Molecular Formula: C15H31NO6
Formula Weight: 321.41
Physical Form: Powder
Appearance: White
Melting Point: 80-90°
Beilstein Reference: 5816747
Optical Rotation: -18° (c=2 in water)
MDL No.: MFCD00134152
Non-ionic detergent for solubilizing membrane proteins
Formel:
C₁₅H₃₁NO₆ Molecular Weight: 321.4 g/mol Schmelzpunkt: 80…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134152 CAS-Nummer: 85316-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...