D-erythro-Sphingosine ≥99%

Lieferant: Thermo Scientific

Synonyme: D-Sphingosine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol, (2S,3R,E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, (2S,3R,E)-2-Aminooctadec-4-ene-1,3-diol

J63584.MB
J63584.MBEA 210 EUR
J63584.MB
D-erythro-Sphingosine ≥99%
D-erythro-Sphingosine

D-erythro-Sphingosine is used to inhibit protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.


Purity: 99+% CAS No.: 123-78-4 Molecular Formula: C18H37NO2 Formula Weight: 299.50 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Beilstein Reference: 1727294 MDL No.: MFCD00036751


Applications: Inhibits protein kinase C and calmodulin-dependent enzymes. Induces apoptosis. A constituent of cell membranes


Bitte beachten Store at –20 °C.

Formel: C₁₈H₃₇NO₂
Molecular Weight: 299,50 g/mol
MDL: MFCD00036751
CAS-Nummer: 123-78-4
EINECS: 204-651-3
Merck Index: 14,8747

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR