5-Thio-D-glucose ≥97%
Lieferant: Thermo Scientific
J63621.MB
J63621.MBEA
128
EUR
J63621.MB
5-Thio-D-glucose ≥97%
5-Thio-D-glucose
Purity: 97+%
CAS No.: 20408-97-3
Molecular Formula: C6H12O5S
Formula Weight: 196.22
Physical Form: Powder
Appearance: White
Melting Point: 135-138°
Beilstein Reference: 1865193
MDL No.: MFCD00006663
Formel:
C₆H₁₂O₅S Molecular Weight: 196,22 g/mol Schmelzpunkt: 135…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006663 CAS-Nummer: 20408-97-3 EINECS: 243-798-8 Merck Index: 14,9334 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...