TAE-Puffer-Lösung 10fach konzentriert (TRIS-Acetat-EDTA-Puffer)

Lieferant: Thermo Fisher Scientific

J63677.K2 J63677.K3 J63677.K7
J63677.K2EA 54.8 EUR
J63677.K2 J63677.K3 J63677.K7
TAE-Puffer-Lösung 10fach konzentriert (TRIS-Acetat-EDTA-Puffer)
Puffer Elektrophorese-Puffer Laufpuffer

Physical Form: Liquid
MDL No.: MFCD00236357

Bestellen

TECHNISCHE DATEN

Environmentally Preferable
Physical form Liquid
MDL No. MFCD00236357

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR