Ambroxol hydrochloride 98%

Lieferant: Thermo Fisher Scientific

Synonyme: trans-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride, Ambroxol HCl, trans-4-((2-Amino-3,5-dibromobenzyl)amino)cyclohexanol hydrochloride

J63712.06
J63712.06EA 165 EUR
J63712.06
Ambroxol hydrochloride 98%
Ambroxol hydrochloride

Purity: 99% CAS No.: 18683-91-5 Molecular Formula: C13H18Br2N2O.HCl Formula Weight: 414.57 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00078932


Applications: Sodium channel blocker. Metabolite of bromhexine

Formel: C₁₃H₁₈Br₂N₂O.HCl
MDL: MFCD00078932
CAS-Nummer: 23828-92-4
EINECS: 242-500-3
Merck Index: 14,386

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR