L(-)-Glutathion (oxidierte Form)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
GSSG, Bi(glutathion-S-yl), L(-)-Glutathion, oxidiert
J63715.03
J63715.06
J63715.03EA
78.1
EUR
J63715.03
J63715.06
L(-)-Glutathion (oxidierte Form)
L(-)-Glutathion (oxidierte Form)
CAS No.: 27025-41-8
Molecular Formula: C20H32N6O12S2
Formula Weight: 612.64
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 1718700
Merck Reference: 14,4475
Optical Rotation: -105° (c=2 in water)
MDL No.: MFCD00063106
Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH
Formel:
C₂₀H₃₂N₆O₁₂S₂ Molecular Weight: 612.64 g/mol Schmelzpunkt: 185 Lagertemperatur: Kühlschrank |
MDL:
MFCD00063106 CAS-Nummer: 27025-41-8 EINECS: 248-170-7 Merck Index: 14,4475 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...