L(-)-Glutathion (oxidierte Form)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: GSSG, Bi(glutathion-S-yl), L(-)-Glutathion, oxidiert

J63715.03 J63715.06
J63715.03EA 78.1 EUR
J63715.03 J63715.06
L(-)-Glutathion (oxidierte Form)
L(-)-Glutathion (oxidierte Form)

CAS No.: 27025-41-8 Molecular Formula: C20H32N6O12S2 Formula Weight: 612.64 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Beilstein Reference: 1718700 Merck Reference: 14,4475 Optical Rotation: -105° (c=2 in water) MDL No.: MFCD00063106


Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH

Formel: C₂₀H₃₂N₆O₁₂S₂
Molecular Weight: 612.64 g/mol
Schmelzpunkt: 185
Lagertemperatur: Kühlschrank
MDL: MFCD00063106
CAS-Nummer: 27025-41-8
EINECS: 248-170-7
Merck Index: 14,4475

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR