Daidzein ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-on, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyron, 4',7-Dihydroxyisoflavon, Daidzeol
J63763.ME
J63763.03
J63763.MC
J63763.MEEA
112
EUR
J63763.ME
J63763.03
J63763.MC
Daidzein ≥98%
Daidzein
Purity: 98+%
CAS No.: 486-66-8
Molecular Formula: C15H10O4
Formula Weight: 254.24
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Melting Point: 328-332° dec.
Beilstein Reference: 231523
Merck Reference: 14,2801
MDL No.: MFCD00016954
Analog of genistein and an agonist at estrogen recpetors
Formel:
C₁₅H₁₀O₄ Siedepunkt: 513 °C (1013 hPa) Schmelzpunkt: 328…332 °C (Zersetzung) |
MDL:
MFCD00016954 CAS-Nummer: 486-66-8 EINECS: 207-635-4 Merck Index: 14,2801 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...