DEAE-Dextran, Pulver

Lieferant: Thermo Scientific

Synonyme: Diethylaminoethyldextran

J63781.14
J63781.14EA 61 EUR
J63781.14
DEAE-Dextran, Pulver
DEAE-Dextran

DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems.


It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.


Bitte beachten Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidising agents.

Lagertemperatur: Raumtemperatur
MDL: MFCD00081561
CAS-Nummer: 9015-73-0
Merck Index: 14,2936

Bestellen

Spezifikation Testergebnisse

Form Powder
Free DEAE-Cl <20 ppm
Loss on Drying <10%
Molecular Weight (viscosity calc.) 500,000 ± 20%
Total Nitrogen 3.0 - 3.8%
Optical purity NMT 0.2 absorption at 375 nm
pH (aqueous solution) 5.0 to 7.0
Sulfated Ash <2%
Viscosity 0.4 to 0.6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR