TEA, 0,2 M buffer soln., pH 8,0
Lieferant: Thermo Fisher Scientific
J63793.AK
J63793.AKEA
95.8
EUR
J63793.AK
TEA, 0,2 M buffer soln., pH 8,0
Puffer
Biologische Puffer
CAS No.: 102-71-6
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002855
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...