2-Chloradenosin

Lieferant: Thermo Fisher Scientific

Synonyme: (2R,3R,4S,5R)-2-(6-Amino-2-chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

J63810.MD J63810.03
J63810.MDEA 121 EUR
J63810.MD J63810.03
2-Chloradenosin
2-Chloradenosin

CAS No.: 146-77-0 Molecular Formula: C10H12ClN5O4 Formula Weight: 301.69 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Beilstein Reference: 43957 MDL No.: MFCD00005734


A selective A1-adenosine receptor agonist. Induces apoptosis

Formel: C₁₀H₁₂ClN₅O₄
Molecular Weight: 301.69 g/mol
MDL: MFCD00005734
CAS-Nummer: 146-77-0
EINECS: 205-678-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR