3,5-Dimethyl-1-adamantanamine hydrochloride
Lieferant: Thermo Fisher Scientific
Synonyme:
Memantine Hydrochloride, 1-Amino-3,5-dimethyladamantane Hydrochloride
J63830.MF
J63830.MFEA
139
EUR
J63830.MF
3,5-Dimethyl-1-adamantanamine hydrochloride
3,5-Dimethyl-1-adamantanamine hydrochloride
CAS No.: 41100-52-1
Molecular Formula: C12H21N.HCl
Formula Weight: 215.76
Physical Form: Powder
Appearance: White
MDL No.: MFCD00214336
Applications: An NMDA receptor antagonist; stimulates dopamine release
Formel:
C₁₂H₂₁N Lagertemperatur: Raumtemperatur |
MDL:
MFCD00214336 CAS-Nummer: 41100-52-1 EINECS: 255-219-6 Merck Index: 14,5829 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...