Proadifen hydrochloride

Lieferant: Thermo Scientific
Achtung

Synonyme: Proadifen HCl

J63833.03
J63833.03EA 342 EUR
J63833.03
Proadifen hydrochloride
Proadifen hydrochloride

CAS No.: 62-68-0 Molecular Formula: C23H31NO2.HCl Formula Weight: 389.97 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Off-white Melting Point: 122-123° Solubility: Soluble in methanol or water MDL No.: MFCD00055151


Applications: Inhibits cytochrome P-450, blocks glibenclamide-sensitive potassium channels. Also inhibits neuronal nitric oxide synthase

Formel: C₂₃H₃₁NO₂·HCl
Molecular Weight: 389,964 g/mol
Schmelzpunkt: 122…123 °C
MDL: MFCD00055151
CAS-Nummer: 62-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR