Methoxychlor

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,1-Bis(4-methoxyphenyl)-2,2,2-trichloroethane, Dimethoxydiphenyltrichloroethane, DMDT

J63839.06
J63839.06EA 43.4 EUR
J63839.06
Methoxychlor
Methoxychlor

CAS No.: 72-43-5 Molecular Formula: C16H15Cl3O2 Formula Weight: 345.65 Physical Form: Crystalline powder Appearance: Off-white Melting Point: 87° Beilstein Reference: 2057367 UN No.: UN3077 MDL No.: MFCD00000803


Applications: Molecule with estrogenic activity

Formel: C₁₆H₁₅Cl₃O₂
Molecular Weight: 345.64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000803
CAS-Nummer: 72-43-5
EINECS: 200-779-9
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR