Apramycin sulphate 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(2R,3R,4S,5S,6S)-2-[[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy-4-hydroxy-3-(methylamino)-2,3,4,4a,6,7,8,8a-octahydropyrano[3,2-b]pyran-2-yl]oxy]-5-amino-6-(hydroxymethyl)oxane-3,4-diol, sulphuric acid, Nebramycin II, Apramycin sulphate salt, (2R,3R,4S,5S,6S)-5-Amino-2-(((2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-(((1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl)oxy)-4-hydroxy-3-(methylamino)octahydropyrano[3,2-b]pyran-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol xsulphate, Nebramycin II sulphate
Purity: 96%
CAS No.: 65710-07-8
Molecular Formula: C21H41N5O11.H2SO4
Formula Weight: 637.65
Storage Temperature: 2°C to 8°C
Flash Point: 451.6°(845°F)
Physical Form: Powder
Density: 1.56
Appearance: White to pale yellow.
Melting Point: 168°
Boiling Point: 823°
Solubility: Soluble in water at 25mg/ml
MDL No.: MFCD06200257
Applications: Potent inhibitor of protein synthesis in bacteria
Formel:
C₂₁H₄₁N₅O₁₁·H₂SO₄ Molecular Weight: 637.65 g/mol Siedepunkt: 823 °C Dichte: 1.56 Flammpunkt: 451.6 °C (845 °F) |
MDL:
MFCD06200257 CAS-Nummer: 65710-07-8 EINECS: 265-890-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...