Factor XA protease digestion buffer

Lieferant: Thermo Fisher Scientific

J63878.AK
J63878.AKEA 58.2 EUR
J63878.AK
Factor XA protease digestion buffer
Puffer Enzympuffer

Physical Form: Liquid
Applications: For His-tag protein purification from mammalian cells

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR