Buffers, Sodium Citrate, 0,5M Solutions

Lieferant: Thermo Fisher Scientific

J62918.AK J62918.AP J61815.AK J61815.AP J63888.AK J63888.AP J63199.AK J63199.AP
J62918.AKEA 70.8 EUR
J62918.AK J62918.AP J61815.AK J61815.AP J63888.AK J63888.AP J63199.AK J63199.AP
Buffers, Sodium Citrate, 0,5M Solutions
Puffer Biologische Puffer

Storage temperature: 2 to 8 °C
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00012462

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR