8-Bromguanosin
Lieferant: Thermo Fisher Scientific
Synonyme:
2-Amino-8-bromo-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one
J63940.06
J63940.06EA
69.5
EUR
J63940.06
8-Bromguanosin
8-Bromguanosin
CAS No.: 4016-63-1
Molecular Formula: C10H12BrN5O5
Formula Weight: 362.14
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD00037985
Formel:
C₁₀H₁₂BrN₅O₅ Molecular Weight: 362.14 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00037985 CAS-Nummer: 4016-63-1 EINECS: 223-677-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...