Safflower oil

Lieferant: Thermo Fisher Scientific

J63982.AP J63982.K2
J63982.APEA 41 EUR
J63982.AP J63982.K2
Safflower oil
Safflower oil

CAS No.: 8001-23-8 Flash Point: 113°(235°F) Physical Form: Oil Density: 0.921 Refractive Index: 1.476 Appearance: Pale yellow MDL No.: MFCD00132216

Dichte: 0,921
Flammpunkt: >113 °C (235 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00132216
CAS-Nummer: 8001-23-8
EINECS: 232-276-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR