2'-Fluoro-2'-deoxyguanosine 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2'-Deoxy-2'-fluoroguanosine, 2′-Deoxy-2′-fluoroguanosine, 2-Amino-9-((2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one
J64055.03
J64055.03EA
101
EUR
J64055.03
2'-Fluoro-2'-deoxyguanosine 99%
2'-Fluoro-2'-deoxyguanosine
Purity: 99%
CAS No.: 78842-13-4
Molecular Formula: C10H12FN5O4
Formula Weight: 285.23
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00923832
Formel:
C₁₀H₁₂FN₅O₄ Molecular Weight: 285,235 g/mol |
MDL:
MFCD00923832 CAS-Nummer: 78842-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...