N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(2R,3R,4R,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-fluorotetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite, Dmt-2'fluoro-da(bz) amidite
J64064.03
J64064.03EA
117
EUR
J64064.03
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite)
Purity: 98%
Molecular Formula: C47H51FN7O7P
Formula Weight: 875.92
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD12912392
Formel:
C₄₇H₅₁FN₇O₇P Molecular Weight: 875,92 g/mol |
MDL:
MFCD12912392 CAS-Nummer: 136834-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...