N-Acetyl-Arg-Glu-Lys-Arg-7-amino-4-(trifluoromethyl)coumarin

Lieferant: Thermo Scientific

J64106.MA
J64106.MAEA 185 EUR
J64106.MA
N-Acetyl-Arg-Glu-Lys-Arg-7-amino-4-(trifluoromethyl)coumarin
N-Acetyl-Arg-Glu-Lys-Arg-7-amino-4-(trifluoromethyl)coumarin

Molecular Formula: C35H51F3N12O9 Formula Weight: 840.80 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Light Sensitive Appearance: White Solubility: Soluble in DMSO or DMF. MDL No.: MFCD03452896


Applications: A fluorogenic substrate suitable for the assay of furin

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR