4-Chlor-N-(propylcarbamoyl)benzolsulfonamid

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Chloro-N-(propylcarbamoyl)benzenesulphonamide, Chlorpropamide

J64110.22
J64110.22EA 222 EUR
J64110.22
4-Chlor-N-(propylcarbamoyl)benzolsulfonamid
1-(4-Chlorophenylsulphonyl)-3-propylurea

CAS No.: 94-20-2 Molecular Formula: C10H13ClN2O3S Formula Weight: 276.74 Physical Form: Powder Appearance: White MDL No.: MFCD00079004


Applications: Increases insulin secretion

Formel: C₁₀H₁₃ClN₂O₃S
Molecular Weight: 276,74 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00079004
CAS-Nummer: 94-20-2
EINECS: 202-314-5
Merck Index: 14,2186

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR