Azadibenzocyclooctyne-PEG4-NHS ester
Lieferant: Thermo Fisher Scientific
Azadibenzocyclooctyne-PEG4-NHS ester (ADIBO-PEG4-NHS ester) is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.
- Form: Oil
- Storage and sensitivity: Store at −20 °C and moisture sensitive
Molecular Formula: C34H39N3O10
Formula Weight: 649.69
Storage Temperature: -30°C to -10°C
Physical Form: Oil
Sensitivity: Moisture Sensitive
Appearance: Orange to brown
Optical Rotation: ADIBO-PEG4-NHS ester
MDL No.: MFCD22380747
Bitte beachten Incompatible with oxidizing agents and heat.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...