1-β-D-Arabinofuranosyluracil 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ara-U, 1-((2R,3S,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, Uracil 1-β-D-Arabinofuranoside
J64147.06
J64147.14
J64147.06EA
41.7
EUR
J64147.06
J64147.14
1-β-D-Arabinofuranosyluracil 99%
1-beta-D-Arabinofuranosyluracil
Purity: 99%
CAS No.: 3083-77-0
Molecular Formula: C9H12N2O6
Formula Weight: 244.20
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Beilstein Reference: 28749
MDL No.: MFCD00065998
Formel:
C₉H₁₂N₂O₆ Molecular Weight: 244.20 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00065998 CAS-Nummer: 3083-77-0 EINECS: 221-386-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...