N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylendiamin

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: TPEN, N1,N1,N2,N2-Tetrakis(pyridin-2-ylmethyl)ethane-1,2-diamine, Tetrakis-(2-pyridylmethyl)ethylenediamine

J64206.MCEA 126 EUR
J64206.MC
N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylendiamin
N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylendiamin

CAS No.: 16858-02-9 Molecular Formula: C26H28N6 Formula Weight: 424.54 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Melting Point: 110-112° Beilstein Reference: 4211982 MDL No.: MFCD00036918


Applications: A cell permeable and extremely high-affinity chelator of heavy metals

Formel: C₂₆H₂₈N₆
Molecular Weight: 424,55 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00036918
CAS-Nummer: 16858-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR