5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine 98%
Lieferant: Thermo Scientific
Synonyme:
1-((2R,3R,4R,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-3-fluoro-4-hydroxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 2'-Deoxy-5'-O-DMT-2'-fluorouridine
J64217.06
J64217.06EA
180
EUR
J64217.06
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine 98%
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine
Purity: 98%
CAS No.: 146954-74-7
Molecular Formula: C30H29N2O7F
Formula Weight: 548.56
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
MDL No.: MFCD06657648
Formel:
C₃₀H₂₉N₂O₇F Molecular Weight: 548,56 g/mol |
MDL:
MFCD06657648 CAS-Nummer: 146954-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...