5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine 98%
Lieferant: Thermo Scientific
J64232.03
J64232.03EA
134
EUR
J64232.03
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine 98%
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine
Purity: 98%
Molecular Formula: C35H36FN5O7
Formula Weight: 657.69
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
MDL No.: MFCD15145247
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...