2-Amino-2'-deoxyadenosine 99%

Lieferant: Thermo Scientific

Synonyme: (2R,3S,5R)-5-(2,6-Diamino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol

J64266.14
J64266.14EA 133 EUR
J64266.14
2-Amino-2'-deoxyadenosine 99%
2-Amino-2'-deoxyadenosine

Purity: 99% CAS No.: 4546-70-7 Molecular Formula: C10H14N6O3 Formula Weight: 266.26 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00047240

Formel: C₁₀H₁₄N₆O₃
Molecular Weight: 266,261 g/mol
MDL: MFCD00047240
CAS-Nummer: 4546-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR